Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCNN1A
Entrez Gene ID
6337
Gene Name
sodium channel epithelial 1 alpha subunit
Gene Aliases
BESC2, ENaCa, ENaCalpha, SCNEA, SCNN1
Chromosome Location
Chr.12: 6346843 - 6377357 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038.5 NP_001029.1 11 - 12 1844 85
NM_001159575.1 NP_001153047.1 11 - 12 1715 85
NM_001159576.1 NP_001153048.1 10 - 11 1996 85
GenBank mRNA AK025536.1 - 6 - 7 568 85
AK172792.1 - 10 - 11 1999 85
AK304379.1 - 11 - 12 1717 85
BC006526.2 - 10 - 11 1682 85
BC062613.1 - 10 - 11 1611 85
DQ402522.1 - 10 - 11 1555 85
X76180.1 - 11 - 12 1654 85

View Full Product Details