The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038.5 | NP_001029.1 | 3 - 4 | 976 | 66 |
NM_001159575.1 | NP_001153047.1 | 3 - 4 | 847 | 66 | |
NM_001159576.1 | NP_001153048.1 | 2 - 3 | 1128 | 66 | |
GenBank mRNA | AK172792.1 | - | 2 - 3 | 1131 | 66 |
AK304379.1 | - | 3 - 4 | 849 | 66 | |
BC006526.2 | - | 2 - 3 | 814 | 66 | |
BC062613.1 | - | 2 - 3 | 743 | 66 | |
DQ402522.1 | - | 2 - 3 | 687 | 66 | |
X76180.1 | - | 3 - 4 | 786 | 66 |