The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003948.3 | NP_003939.1 | 8 - 9 | 1540 | 158 |
XM_005263335.2 | XP_005263392.1 | 8 - 9 | 1259 | 158 | |
XM_006714406.2 | XP_006714469.1 | 8 - 9 | 1259 | 158 | |
XM_017008809.1 | XP_016864298.1 | 8 - 9 | 1259 | 158 | |
XM_017008810.1 | XP_016864299.1 | 8 - 9 | 1259 | 158 | |
XM_017008811.1 | XP_016864300.1 | 8 - 9 | 1259 | 158 | |
GenBank mRNA | AK294866.1 | - | 8 - 9 | 1526 | 158 |
AK309127.1 | - | 1 - 2 | 82 | 158 | |
AK312490.1 | - | 8 - 9 | 1531 | 158 | |
BC093646.1 | - | 8 - 9 | 1064 | 158 | |
BC093981.1 | - | 8 - 9 | 1064 | 158 | |
U35146.1 | - | 8 - 9 | 1390 | 158 |