The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021133.3 | NP_066956.1 | 5 - 6 | 2154 | 129 |
GenBank mRNA | AK315231.1 | - | 5 - 6 | 2105 | 130 |
BC114433.1 | - | 4 - 5 | 2020 | 129 | |
BC115697.1 | - | 4 - 5 | 2020 | 129 |