The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020474.3 | NP_065207.2 | 3 - 4 | 570 | 139 |
XM_005258239.3 | XP_005258296.1 | 4 - 5 | 638 | 139 | |
XM_017025692.1 | XP_016881181.1 | 4 - 5 | 817 | 139 | |
GenBank mRNA | AK293062.1 | - | 5 - 6 | 712 | 139 |
BC038440.1 | - | 4 - 5 | 635 | 139 | |
DN996147.1 | - | 3 - 4 | 588 | 139 | |
U41514.1 | - | 3 - 4 | 507 | 139 | |
X85018.1 | - | 3 - 4 | 476 | 139 |