The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300905.1 | NP_001287834.1 | 16 - 17 | 3248 | 81 |
NM_013449.3 | NP_038477.2 | 16 - 17 | 3239 | 81 | |
XM_011537797.2 | XP_011536099.1 | 16 - 17 | 3153 | 81 | |
XM_011537798.2 | XP_011536100.1 | 16 - 17 | 3153 | 81 | |
XM_011537799.2 | XP_011536101.1 | 17 - 18 | 3063 | 81 | |
XM_011537800.2 | XP_011536102.1 | 16 - 17 | 3205 | 81 | |
XM_011537801.2 | XP_011536103.1 | 16 - 17 | 3232 | 81 | |
XM_011537803.2 | XP_011536105.1 | 16 - 17 | 3248 | 81 | |
XM_011537804.2 | XP_011536106.1 | 18 - 19 | 3331 | 81 | |
XM_011537805.2 | XP_011536107.1 | 17 - 18 | 3334 | 81 | |
XM_011537806.2 | XP_011536108.1 | 16 - 17 | 3112 | 81 | |
XM_011537807.2 | XP_011536109.1 | 15 - 16 | 2919 | 81 | |
XM_011537809.2 | XP_011536111.1 | 10 - 11 | 1452 | 81 | |
XM_017018740.1 | XP_016874229.1 | 17 - 18 | 3379 | 81 | |
XM_017018741.1 | XP_016874230.1 | 17 - 18 | 3153 | 81 | |
XM_017018742.1 | XP_016874231.1 | 18 - 19 | 3289 | 81 | |
GenBank mRNA | AB002312.2 | - | 15 - 16 | 3028 | 81 |
AK023830.1 | - | 14 - 15 | 2874 | 81 | |
AK023842.1 | - | 16 - 17 | 3127 | 81 | |
CR749379.1 | - | 16 - 17 | 3240 | 81 |