The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300905.1 | NP_001287834.1 | 4 - 5 | 1126 | 73 |
NM_013449.3 | NP_038477.2 | 4 - 5 | 1117 | 73 | |
XM_011537797.2 | XP_011536099.1 | 4 - 5 | 1031 | 73 | |
XM_011537798.2 | XP_011536100.1 | 4 - 5 | 1031 | 73 | |
XM_011537799.2 | XP_011536101.1 | 5 - 6 | 941 | 73 | |
XM_011537800.2 | XP_011536102.1 | 4 - 5 | 1083 | 73 | |
XM_011537801.2 | XP_011536103.1 | 4 - 5 | 1110 | 73 | |
XM_011537803.2 | XP_011536105.1 | 4 - 5 | 1126 | 73 | |
XM_011537804.2 | XP_011536106.1 | 6 - 7 | 1209 | 73 | |
XM_011537805.2 | XP_011536107.1 | 5 - 6 | 1212 | 73 | |
XM_011537806.2 | XP_011536108.1 | 4 - 5 | 990 | 73 | |
XM_011537807.2 | XP_011536109.1 | 4 - 5 | 1031 | 73 | |
XM_017018740.1 | XP_016874229.1 | 5 - 6 | 1257 | 73 | |
XM_017018741.1 | XP_016874230.1 | 5 - 6 | 1031 | 73 | |
XM_017018742.1 | XP_016874231.1 | 6 - 7 | 1167 | 73 | |
GenBank mRNA | AB002312.2 | - | 3 - 4 | 906 | 73 |
AK023830.1 | - | 2 - 3 | 752 | 73 | |
AK023842.1 | - | 4 - 5 | 1006 | 73 | |
CR749379.1 | - | 4 - 5 | 1118 | 73 |