The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286124.1 | NP_001273053.1 | 7 - 8 | 1191 | 151 |
NM_001286126.1 | NP_001273055.1 | 7 - 8 | 1191 | 151 | |
NM_001286128.1 | NP_001273057.1 | 6 - 7 | 1125 | 151 | |
NM_018650.4 | NP_061120.3 | 7 - 8 | 1191 | 151 | |
XM_005273134.4 | XP_005273191.1 | 7 - 8 | 1058 | 151 | |
XM_006711326.3 | XP_006711389.1 | 7 - 8 | 452 | 151 | |
XM_011509561.2 | XP_011507863.1 | 7 - 8 | 1187 | 151 | |
XM_011509562.2 | XP_011507864.1 | 7 - 8 | 804 | 151 | |
XM_017001305.1 | XP_016856794.1 | 8 - 9 | 648 | 151 | |
XM_017001306.1 | XP_016856795.1 | 7 - 8 | 688 | 151 | |
XM_017001307.1 | XP_016856796.1 | 8 - 9 | 1088 | 151 | |
GenBank mRNA | AB040910.1 | - | 6 - 7 | 1082 | 151 |
AF154845.1 | - | 7 - 8 | 817 | 151 | |
AK027493.1 | - | 3 - 4 | 148 | 151 | |
AK295506.1 | - | 7 - 8 | 689 | 151 | |
AK314614.1 | - | 7 - 8 | 851 | 151 | |
BC113869.1 | - | 7 - 8 | 727 | 151 | |
BC114478.1 | - | 7 - 8 | 727 | 151 | |
KJ534879.1 | - | 7 - 8 | 551 | 151 | |
KJ534881.1 | - | 7 - 8 | 551 | 151 | |
KJ535089.1 | - | 7 - 8 | 550 | 151 |