The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317231.1 | NP_001304160.1 | 12 - 13 | 1018 | 128 |
NM_001317232.1 | NP_001304161.1 | 11 - 12 | 991 | 128 | |
NM_001317233.1 | NP_001304162.1 | 11 - 12 | 960 | 128 | |
NM_014342.3 | NP_055157.1 | 12 - 13 | 1018 | 128 | |
XM_006718172.2 | XP_006718235.1 | 11 - 12 | 893 | 128 | |
XM_017017462.1 | XP_016872951.1 | 10 - 11 | 913 | 128 | |
XM_017017463.1 | XP_016872952.1 | 12 - 13 | 937 | 128 | |
GenBank mRNA | AF085361.1 | - | 12 - 13 | 877 | 128 |
AF176008.1 | - | 12 - 13 | 878 | 128 | |
AK295218.1 | - | 11 - 12 | 840 | 128 | |
AY380792.1 | - | 12 - 13 | 876 | 128 | |
BC000875.1 | - | 12 - 13 | 874 | 128 |