The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018649.2 | NP_061119.1 | 5 - 6 | 1052 | 87 |
GenBank mRNA | AF151534.1 | - | 5 - 6 | 806 | 87 |
AF336304.1 | - | 5 - 6 | 784 | 87 | |
AK022475.1 | - | 5 - 6 | 776 | 87 | |
AK022776.1 | - | 5 - 6 | 807 | 87 | |
AK131057.1 | - | 5 - 6 | 803 | 87 | |
BC016172.2 | - | 5 - 6 | 808 | 87 |