Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNK1
Entrez Gene ID
65125
Gene Name
WNK lysine deficient protein kinase 1
Gene Aliases
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
Chromosome Location
Chr.12: 752552 - 911452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184985.1 NP_001171914.1 24 - 25 7663 92
NM_014823.2 NP_055638.2 22 - 23 6139 92
NM_018979.3 NP_061852.3 24 - 25 6883 92
NM_213655.4 NP_998820.3 24 - 25 7639 92
XM_006719003.2 XP_006719066.1 24 - 25 7205 92
XM_011520997.2 XP_011519299.1 25 - 26 8493 92
XM_011520998.2 XP_011519300.1 25 - 26 8444 92
XM_011520999.2 XP_011519301.1 25 - 26 8444 92
XM_011521000.2 XP_011519302.1 25 - 26 8447 92
XM_011521001.2 XP_011519303.1 24 - 25 8168 92
XM_011521002.2 XP_011519304.1 24 - 25 7985 92
XM_011521003.2 XP_011519305.1 23 - 24 7709 92
XM_011521004.2 XP_011519306.1 23 - 24 7706 92
XM_011521005.2 XP_011519307.1 22 - 23 6931 92
XM_011521006.2 XP_011519308.1 23 - 24 7124 92
XM_011521007.2 XP_011519309.1 23 - 24 7121 92
XM_011521008.2 XP_011519310.1 21 - 22 6386 92
XM_011521009.2 XP_011519311.1 21 - 22 6383 92
XM_017019834.1 XP_016875323.1 23 - 24 6746 92
XM_017019835.1 XP_016875324.1 22 - 23 6665 92
XM_017019836.1 XP_016875325.1 22 - 23 6662 92
XM_017019837.1 XP_016875326.1 22 - 23 6470 92
XM_017019838.1 XP_016875327.1 22 - 23 6467 92
XM_017019839.1 XP_016875328.1 21 - 22 6386 92
GenBank mRNA AB002342.2 - 22 - 23 5294 92
AJ296290.1 - 24 - 25 6240 92
BC141881.1 - 16 - 17 3707 92
JQ358908.1 - 25 - 26 7479 92

View Full Product Details