The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005181.3 | NP_005172.1 | 1 - 2 | 116 | 79 |
GenBank mRNA | AK096880.1 | - | 1 - 2 | 116 | 79 |
AK300670.1 | - | 1 - 2 | 85 | 79 | |
AK300677.1 | - | - | 85 | 79 | |
AK313254.1 | - | 1 - 2 | 85 | 79 | |
BC004897.2 | - | 1 - 2 | 71 | 79 | |
DA421884.1 | - | 1 - 2 | 116 | 79 | |
EU794596.1 | - | 1 - 2 | 716 | 79 |