The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184957.1 | NP_001171886.1 | 13 - 14 | 2193 | 83 |
NM_001184958.1 | NP_001171887.1 | 13 - 14 | 2298 | 83 | |
NM_032137.4 | NP_115513.4 | 13 - 14 | 2695 | 83 | |
XM_006713341.2 | XP_006713404.1 | 13 - 14 | 2695 | 83 | |
XM_006713342.2 | XP_006713405.1 | 13 - 14 | 2548 | 83 | |
XM_011534153.2 | XP_011532455.1 | 13 - 14 | 4325 | 83 | |
XM_011534154.2 | XP_011532456.1 | 13 - 14 | 3760 | 83 | |
XM_011534157.2 | XP_011532459.1 | 9 - 10 | 1472 | 83 | |
GenBank mRNA | AK131481.1 | - | 14 - 15 | 2901 | 83 |
AL136781.1 | - | 13 - 14 | 2113 | 83 | |
AL834386.1 | - | 13 - 14 | 2647 | 83 | |
BC030599.2 | - | 13 - 14 | 2256 | 83 | |
BC038406.1 | - | 13 - 14 | 2615 | 83 |