Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3orf20
Entrez Gene ID
84077
Gene Name
chromosome 3 open reading frame 20
Gene Aliases
-
Chromosome Location
Chr.3: 14673532 - 14773036 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184957.1 NP_001171886.1 4 - 5 579 69
NM_001184958.1 NP_001171887.1 4 - 5 684 69
NM_032137.4 NP_115513.4 4 - 5 1081 69
XM_006713341.2 XP_006713404.1 4 - 5 1081 69
XM_006713342.2 XP_006713405.1 4 - 5 934 69
XM_006713343.2 XP_006713406.1 4 - 5 1081 69
XM_011534153.2 XP_011532455.1 4 - 5 2711 69
XM_011534154.2 XP_011532456.1 4 - 5 2146 69
XM_011534158.2 XP_011532460.1 4 - 5 1081 69
XM_017007310.1 XP_016862799.1 4 - 5 1081 69
GenBank mRNA AK131481.1 - 4 - 5 935 69
AL136781.1 - 4 - 5 499 69
AL834386.1 - 4 - 5 1033 69
BC030599.2 - 4 - 5 642 69
BC038406.1 - 4 - 5 1001 69

View Full Product Details