The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207420.2 | NP_997303.2 | 11 - 12 | 1319 | 77 |
GenBank mRNA | AY571960.2 | - | 11 - 12 | 1319 | 77 |