The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207420.2 | NP_997303.2 | 4 - 5 | 436 | 81 |
XM_011540824.2 | XP_011539126.1 | 4 - 5 | 475 | 81 | |
XM_011540825.2 | XP_011539127.1 | 4 - 5 | 475 | 81 | |
GenBank mRNA | AY571960.2 | - | 4 - 5 | 436 | 81 |