The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127895.1 | NP_001121367.1 | 4 - 5 | 672 | 89 |
NM_001127896.1 | NP_001121368.1 | 3 - 4 | 583 | 89 | |
NM_022467.3 | NP_071912.2 | 3 - 4 | 925 | 89 | |
XM_011527222.1 | XP_011525524.1 | 3 - 4 | 906 | 89 | |
XM_011527224.1 | XP_011525526.1 | 4 - 5 | 1040 | 89 | |
XM_017027143.1 | XP_016882632.1 | 3 - 4 | 576 | 89 | |
GenBank mRNA | AB047801.1 | - | 3 - 4 | 439 | 89 |
AF300612.1 | - | 3 - 4 | 386 | 89 | |
AF305781.1 | - | 3 - 4 | 482 | 89 | |
BC011380.1 | - | 3 - 4 | 535 | 89 | |
BC014250.2 | - | 3 - 4 | 579 | 89 | |
BC018723.1 | - | 3 - 4 | 925 | 89 | |
BI917117.1 | - | 3 - 4 | 583 | 89 | |
BX354555.2 | - | 4 - 5 | 671 | 89 |