Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB45
Entrez Gene ID
84878
Gene Name
zinc finger and BTB domain containing 45
Gene Aliases
ZNF499
Chromosome Location
Chr.19: 58513530 - 58541712 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316979.1 NP_001303908.1 1 - 2 268 74
GenBank mRNA AK027392.1 - 1 - 2 80 74
AK130056.1 - 1 - 2 69 74
HY070267.1 - 1 - 2 270 74

View Full Product Details