The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152544.2 | NP_689757.2 | 4 - 5 | 1041 | 107 |
XM_005247950.3 | XP_005248007.1 | 4 - 5 | 1038 | 107 | |
XM_011513406.2 | XP_011511708.1 | 4 - 5 | 1070 | 107 | |
XM_011513407.2 | XP_011511709.1 | 4 - 5 | 1070 | 107 | |
XM_011513408.1 | XP_011511710.1 | 5 - 6 | 1182 | 107 | |
XM_011513409.2 | XP_011511711.1 | 5 - 6 | 1153 | 107 | |
XM_011513410.2 | XP_011511712.1 | 4 - 5 | 1072 | 107 | |
XM_017007811.1 | XP_016863300.1 | 4 - 5 | 1071 | 107 | |
GenBank mRNA | BC035655.1 | - | 4 - 5 | 541 | 107 |