The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182543.3 | NP_872349.1 | 2 - 3 | 666 | 143 |
XM_011519384.1 | XP_011517686.1 | 4 - 5 | 638 | 143 | |
XM_011519385.1 | XP_011517687.1 | 3 - 4 | 327 | 143 | |
XM_011519386.2 | XP_011517688.1 | 3 - 4 | 380 | 143 | |
XM_017015907.1 | XP_016871396.1 | 3 - 4 | 563 | 143 | |
XM_017015908.1 | XP_016871397.1 | 3 - 4 | 380 | 143 | |
GenBank mRNA | AK074323.1 | - | 2 - 3 | 619 | 143 |
BC035778.1 | - | 2 - 3 | 652 | 143 | |
DA332885.1 | - | 3 - 4 | 327 | 143 |