The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284347.1 | NP_001271276.1 | 8 - 9 | 1272 | 124 |
NM_001284348.1 | NP_001271277.1 | 10 - 11 | 1507 | 124 | |
NM_001322470.1 | NP_001309399.1 | 9 - 10 | 1401 | 124 | |
NM_001322471.1 | NP_001309400.1 | 8 - 9 | 1272 | 124 | |
NM_001322473.1 | NP_001309402.1 | 9 - 10 | 1401 | 124 | |
NM_001322475.1 | NP_001309404.1 | 10 - 11 | 1519 | 124 | |
XM_011521912.2 | XP_011520214.1 | 13 - 14 | 1876 | 124 | |
XM_017022493.1 | XP_016877982.1 | 12 - 13 | 1837 | 124 | |
XM_017022495.1 | XP_016877984.1 | 13 - 14 | 1876 | 124 | |
XM_017022496.1 | XP_016877985.1 | 13 - 14 | 1876 | 124 | |
XM_017022497.1 | XP_016877986.1 | 13 - 14 | 1876 | 124 | |
XM_017022499.1 | XP_016877988.1 | 13 - 14 | 1876 | 124 | |
XM_017022502.1 | XP_016877991.1 | 10 - 11 | 1530 | 124 | |
XM_017022504.1 | XP_016877993.1 | 8 - 9 | 1295 | 124 | |
GenBank mRNA | AK022538.1 | - | 10 - 11 | 1457 | 124 |
BC039292.1 | - | 8 - 9 | 1222 | 124 |