The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308295.1 | NP_001295224.1 | 8 - 9 | 1028 | 152 |
NM_004808.2 | NP_004799.1 | 7 - 8 | 971 | 152 | |
XM_005252642.4 | XP_005252699.1 | 7 - 8 | 1007 | 152 | |
XM_017016947.1 | XP_016872436.1 | 7 - 8 | 1007 | 152 | |
XM_017016948.1 | XP_016872437.1 | 8 - 9 | 946 | 152 | |
XM_017016949.1 | XP_016872438.1 | 8 - 9 | 946 | 152 | |
XM_017016952.1 | XP_016872441.1 | 8 - 9 | 1167 | 152 | |
XM_017016953.1 | XP_016872442.1 | 8 - 9 | 1167 | 152 | |
GenBank mRNA | AF043325.1 | - | 7 - 8 | 933 | 152 |
AK295277.1 | - | 8 - 9 | 907 | 152 | |
AK302101.1 | - | 7 - 8 | 1048 | 152 | |
AK315085.1 | - | 7 - 8 | 899 | 152 | |
BC006376.2 | - | 7 - 8 | 889 | 152 | |
BT007024.1 | - | 7 - 8 | 887 | 152 |