The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317184.1 | NP_001304113.1 | 14 - 15 | 2386 | 110 |
NM_001317185.1 | NP_001304114.1 | 15 - 16 | 2636 | 110 | |
NM_001317186.1 | NP_001304115.1 | 14 - 15 | 2423 | 110 | |
NM_004360.4 | NP_004351.1 | 15 - 16 | 2569 | 110 | |
XM_011523488.1 | XP_011521790.1 | 14 - 15 | 2433 | 110 | |
XM_011523489.1 | XP_011521791.1 | 14 - 15 | 2720 | 110 | |
GenBank mRNA | AB025105.1 | - | 14 - 15 | 2324 | 110 |
AB025106.1 | - | 15 - 16 | 2564 | 110 | |
AK290012.1 | - | 15 - 16 | 2541 | 110 | |
AK297913.1 | - | 11 - 12 | 1692 | 110 | |
AK309703.1 | - | 14 - 15 | 2515 | 110 | |
AK312551.1 | - | 15 - 16 | 2569 | 110 | |
EU709494.1 | - | 15 - 16 | 2567 | 110 | |
L08599.1 | - | 15 - 16 | 2539 | 110 | |
Z13009.1 | - | 15 - 16 | 2537 | 110 | |
Z18923.1 | - | 15 - 16 | 2443 | 110 |