The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005293.2 | NP_005284.2 | 2 - 2 | 1389 | 63 |
GenBank mRNA | AA601037.1 | - | 1 - 1 | 63 | |
AB209066.1 | - | 1 - 1 | 1047 | 63 |