The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000795.3 | NP_000786.1 | 4 - 5 | 769 | 58 |
NM_016574.3 | NP_057658.2 | 4 - 5 | 769 | 58 | |
XM_017017296.1 | XP_016872785.1 | 4 - 5 | 751 | 58 | |
GenBank mRNA | AB209832.1 | - | 3 - 4 | 564 | 58 |
AB451480.1 | - | 3 - 4 | 534 | 58 | |
AF176812.1 | - | 3 - 4 | 543 | 58 | |
AK308777.1 | - | 4 - 5 | 599 | 58 | |
BC021195.2 | - | 4 - 5 | 629 | 58 | |
M29066.1 | - | 3 - 4 | 567 | 58 | |
M30625.1 | - | 4 - 5 | 870 | 58 | |
S62137.1 | - | 3 - 4 | 851 | 58 | |
S69899.1 | - | 4 - 5 | 633 | 58 | |
X51362.1 | - | 4 - 5 | 699 | 58 | |
X51645.1 | - | 3 - 4 | 562 | 58 |