The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301108.1 | NP_001288037.1 | 6 - 7 | 925 | 113 |
NM_001301109.1 | NP_001288038.1 | 9 - 10 | 1445 | 113 | |
NM_001324182.1 | NP_001311111.1 | 6 - 7 | 932 | 113 | |
NM_001324183.1 | NP_001311112.1 | 7 - 8 | 1083 | 113 | |
NM_001324184.1 | NP_001311113.1 | 8 - 9 | 1414 | 113 | |
NM_001324185.1 | NP_001311114.1 | 7 - 8 | 1076 | 113 | |
NM_001324186.1 | NP_001311115.1 | 7 - 8 | 1263 | 113 | |
NM_001324187.1 | NP_001311116.1 | 6 - 7 | 863 | 113 | |
NM_003027.4 | NP_003018.3 | 6 - 7 | 932 | 113 | |
NR_125370.1 | 6 - 7 | 932 | 113 | ||
NR_136711.1 | 7 - 8 | 1076 | 113 | ||
NR_136712.1 | 7 - 8 | 1263 | 113 | ||
NR_136713.1 | 6 - 7 | 925 | 113 | ||
NR_136714.1 | 6 - 7 | 925 | 113 | ||
XM_011521889.1 | XP_011520191.1 | 8 - 9 | 1314 | 113 | |
XM_017022486.1 | XP_016877975.1 | 6 - 7 | 1030 | 113 | |
GenBank mRNA | AF036269.1 | - | 6 - 7 | 629 | 113 |
AF036270.1 | - | 6 - 7 | 622 | 113 | |
AF036271.1 | - | 9 - 10 | 1142 | 113 | |
AK307442.1 | - | 6 - 7 | 777 | 113 | |
BC035598.1 | - | 6 - 7 | 649 | 113 | |
BC042864.1 | - | 6 - 7 | 773 | 113 | |
X99664.1 | - | 6 - 7 | 626 | 113 |