Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBL7
Entrez Gene ID
84993
Gene Name
ubiquitin like 7
Gene Aliases
BMSC-UbP, TCBA1
Chromosome Location
Chr.15: 74445977 - 74461188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286739.1 NP_001273668.1 5 - 6 583 87
NM_001286740.1 NP_001273669.1 4 - 5 448 87
NM_001286741.1 NP_001273670.1 4 - 5 938 87
NM_001286742.1 NP_001273671.1 5 - 6 680 87
NM_032907.4 NP_116296.1 4 - 5 569 87
NM_201265.1 NP_957717.1 4 - 5 498 87
XM_011522124.2 XP_011520426.1 5 - 6 785 87
GenBank mRNA AY037166.1 - 4 - 5 472 87
BC007913.2 - 4 - 5 852 87
BC030055.1 - 5 - 6 546 87
BC033919.1 - 5 - 6 587 87
BG704085.1 - 4 - 5 495 87
BI551787.1 - 4 - 5 555 87
BI552240.1 - 4 - 5 504 87

View Full Product Details