The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286739.1 | NP_001273668.1 | 9 - 10 | 914 | 79 |
NM_001286740.1 | NP_001273669.1 | 8 - 9 | 779 | 79 | |
NM_001286741.1 | NP_001273670.1 | 8 - 9 | 1269 | 79 | |
NM_001286742.1 | NP_001273671.1 | 9 - 10 | 1011 | 79 | |
NM_032907.4 | NP_116296.1 | 8 - 9 | 900 | 79 | |
NM_201265.1 | NP_957717.1 | 8 - 9 | 829 | 79 | |
XM_011522124.2 | XP_011520426.1 | 9 - 10 | 1116 | 79 | |
GenBank mRNA | AY037166.1 | - | 8 - 9 | 803 | 79 |
BC007913.2 | - | 8 - 9 | 1183 | 79 | |
BC030055.1 | - | 9 - 10 | 877 | 79 | |
BC033919.1 | - | 9 - 10 | 918 | 79 |