Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF38
Entrez Gene ID
152006
Gene Name
ring finger protein 38
Gene Aliases
-
Chromosome Location
Chr.9: 36336398 - 36487384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011517712.2 XP_011516014.1 2 - 3 148 74

View Full Product Details