Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMD12
Entrez Gene ID
5718
Gene Name
proteasome 26S subunit, non-ATPase 12
Gene Aliases
Rpn5, p55
Chromosome Location
Chr.17: 67337916 - 67366627 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316341.1 NP_001303270.1 8 - 9 1046 131
NM_002816.4 NP_002807.1 6 - 7 762 131
NM_174871.3 NP_777360.1 5 - 6 702 131
GenBank mRNA AB003103.1 - 6 - 7 697 131
AK091198.1 - 5 - 6 680 131
AK222679.1 - 6 - 7 679 131
AK307821.1 - 6 - 7 712 131
AK308883.1 - 6 - 7 703 131
BC019062.2 - 6 - 7 693 131
BC065826.1 - 8 - 9 966 131
KU178288.1 - 6 - 7 654 131
KU178289.1 - 6 - 7 654 131

View Full Product Details