The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276453.1 | NP_001263382.1 | 16 - 17 | 2186 | 71 |
NM_002265.5 | NP_002256.2 | 17 - 18 | 2511 | 71 | |
GenBank mRNA | AK299145.1 | - | 12 - 13 | 1543 | 71 |
AK301463.1 | - | 16 - 17 | 2186 | 71 | |
AK314780.1 | - | 17 - 18 | 2389 | 71 | |
AK316421.1 | - | 16 - 17 | 2407 | 71 | |
BC003572.1 | - | 17 - 18 | 2385 | 71 | |
BC024045.1 | - | 17 - 18 | 2385 | 71 | |
BC036703.2 | - | 17 - 18 | 2411 | 71 | |
BT009797.1 | - | 17 - 18 | 2104 | 71 | |
L38951.1 | - | 17 - 18 | 2441 | 71 |