Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKAG2
Entrez Gene ID
51422
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 2
Gene Aliases
AAKG, AAKG2, CMH6, H91620p, WPWS
Chromosome Location
Chr.7: 151556114 - 151877231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040633.1 NP_001035723.1 11 - 12 1567 97
NM_001304527.1 NP_001291456.1 9 - 10 1171 97
NM_001304531.1 NP_001291460.1 8 - 9 589 97
NM_016203.3 NP_057287.2 11 - 12 1836 97
NM_024429.1 NP_077747.1 7 - 8 661 97
XM_005250002.3 XP_005250059.1 11 - 12 1702 97
XM_005250004.3 XP_005250061.1 11 - 12 1648 97
XM_005250006.4 XP_005250063.1 9 - 10 1401 97
XM_006716021.2 XP_006716084.1 11 - 12 1239 97
XM_011516282.1 XP_011514584.1 11 - 12 1236 97
XM_011516283.1 XP_011514585.1 11 - 12 1239 97
XM_011516284.1 XP_011514586.1 11 - 12 1236 97
XM_011516285.1 XP_011514587.1 8 - 9 1081 97
XM_011516286.1 XP_011514588.1 7 - 8 602 97
XM_011516287.1 XP_011514589.1 10 - 11 1312 97
XM_017012268.1 XP_016867757.1 11 - 12 1645 97
XM_017012269.1 XP_016867758.1 11 - 12 1450 97
XM_017012270.1 XP_016867759.1 11 - 12 1226 97
XM_017012271.1 XP_016867760.1 11 - 12 1645 97
XM_017012272.1 XP_016867761.1 11 - 12 1223 97
XM_017012273.1 XP_016867762.1 9 - 10 1386 97
XM_017012274.1 XP_016867763.1 8 - 9 1078 97
XM_017012275.1 XP_016867764.1 10 - 11 1575 97
XM_017012276.1 XP_016867765.1 11 - 12 1671 97
XM_017012277.1 XP_016867766.1 7 - 8 602 97
XM_017012278.1 XP_016867767.1 10 - 11 1312 97
XM_017012279.1 XP_016867768.1 10 - 11 1578 97
XM_017012280.1 XP_016867769.1 10 - 11 1575 97
XM_017012281.1 XP_016867770.1 10 - 11 1601 97
XM_017012282.1 XP_016867771.1 9 - 10 1021 97
GenBank mRNA AB025580.1 - 7 - 8 574 97
AF087875.1 - 7 - 8 622 97
AK001887.1 - 7 - 8 661 97
AK301123.1 - 9 - 10 1120 97
AK314646.1 - 7 - 8 623 97
BC020540.1 - 7 - 8 542 97
BC068598.1 - 11 - 12 1536 97
BT007127.1 - 7 - 8 502 97

View Full Product Details