The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014159.6 | NP_054878.5 | 18 - 19 | 7403 | 72 |
XM_011533631.2 | XP_011531933.1 | 19 - 20 | 8899 | 72 | |
XM_011533632.2 | XP_011531934.1 | 18 - 19 | 7475 | 72 | |
XM_011533633.2 | XP_011531935.1 | 18 - 19 | 8766 | 72 | |
XM_011533634.1 | XP_011531936.1 | 18 - 19 | 7428 | 72 | |
XM_017006270.1 | XP_016861759.1 | 16 - 17 | 8602 | 72 | |
GenBank mRNA | AB051519.2 | - | 16 - 17 | 5403 | 72 |
AF049103.1 | - | 2 - 3 | 193 | 72 | |
AF049610.1 | - | 2 - 3 | 221 | 72 | |
AJ238403.1 | - | 16 - 17 | 6250 | 72 | |
AK026837.1 | - | 4 - 5 | 1061 | 72 | |
AK095494.1 | - | 11 - 12 | 1859 | 72 | |
AL713692.1 | - | 4 - 5 | 669 | 72 | |
AL831959.1 | - | 16 - 17 | 6545 | 72 | |
AY576987.1 | - | 16 - 17 | 5912 | 72 | |
AY576988.1 | - | 18 - 19 | 5991 | 72 | |
BC090954.1 | - | 16 - 17 | 5193 | 72 | |
BC117162.1 | - | 16 - 17 | 6026 | 72 | |
BC117164.1 | - | 16 - 17 | 6026 | 72 | |
BX649110.1 | - | 16 - 17 | 7170 | 72 |