The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014159.6 | NP_054878.5 | 11 - 12 | 5441 | 62 |
XM_011533631.2 | XP_011531933.1 | 12 - 13 | 6937 | 62 | |
XM_011533632.2 | XP_011531934.1 | 11 - 12 | 5513 | 62 | |
XM_011533633.2 | XP_011531935.1 | 11 - 12 | 6804 | 62 | |
XM_011533634.1 | XP_011531936.1 | 11 - 12 | 5466 | 62 | |
XM_017006270.1 | XP_016861759.1 | 9 - 10 | 6640 | 62 | |
GenBank mRNA | AB051519.2 | - | 9 - 10 | 3441 | 62 |
AF161554.1 | - | 9 - 10 | 1185 | 62 | |
AL831959.1 | - | 8 - 9 | 4416 | 62 | |
AY576987.1 | - | 9 - 10 | 3950 | 62 | |
AY576988.1 | - | 10 - 11 | 3953 | 62 | |
BC090954.1 | - | 9 - 10 | 3231 | 62 | |
BC117162.1 | - | 9 - 10 | 4064 | 62 | |
BC117164.1 | - | 9 - 10 | 4064 | 62 | |
BX649110.1 | - | 9 - 10 | 5208 | 62 |