Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C16orf45
Entrez Gene ID
89927
Gene Name
chromosome 16 open reading frame 45
Gene Aliases
MINP
Chromosome Location
Chr.16: 15434468 - 15594521 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142469.1 NP_001135941.1 2 - 3 232 74
NM_033201.2 NP_149978.1 2 - 3 413 74
XM_011522731.2 XP_011521033.1 2 - 3 392 74
GenBank mRNA AK057180.1 - 2 - 3 413 74
AK293431.1 - 2 - 3 319 74
BC008967.2 - 2 - 3 343 74
BC023603.2 - 2 - 3 232 74

View Full Product Details