Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SBSN
Entrez Gene ID
374897
Gene Name
suprabasin
Gene Aliases
UNQ698
Chromosome Location
Chr.19: 35523367 - 35528371 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166034.1 NP_001159506.1 1 - 1 1310 70
XM_011526929.2 XP_011525231.1 2 - 2 1276 70
XM_011526930.2 XP_011525232.1 2 - 2 1232 70
XM_011526931.2 XP_011525233.1 2 - 2 1222 70

View Full Product Details