The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024334.2 | NP_077310.1 | 1 - 2 | 261 | 95 |
GenBank mRNA | AK027757.1 | - | 1 - 2 | 187 | 95 |
AK027877.1 | - | 1 - 2 | 125 | 95 | |
AK075010.1 | - | 1 - 2 | 112 | 95 | |
AK225589.1 | - | 1 - 2 | 184 | 95 | |
AL136916.1 | - | 1 - 2 | 155 | 95 | |
BC003125.1 | - | 1 - 2 | 156 | 95 | |
BC011719.2 | - | 1 - 2 | 156 | 95 | |
DA259919.1 | - | 1 - 2 | 266 | 95 |