The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326.2 | NP_001317.1 | 8 - 9 | 754 | 150 |
GenBank mRNA | AK290836.1 | - | 8 - 9 | 754 | 150 |
AK294204.1 | - | 5 - 6 | 347 | 150 | |
AK297371.1 | - | 7 - 8 | 636 | 150 | |
BC108319.1 | - | 8 - 9 | 728 | 150 | |
U15782.1 | - | 8 - 9 | 719 | 150 |