Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCO3
Entrez Gene ID
55002
Gene Name
transmembrane and coiled-coil domains 3
Gene Aliases
C13orf11
Chromosome Location
Chr.13: 113490993 - 113550229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017905.4 NP_060375.4 6 - 7 1393 115
XM_006719969.1 XP_006720032.1 6 - 7 1174 115
XM_011537498.2 XP_011535800.1 6 - 7 1517 115
XM_011537501.2 XP_011535803.1 6 - 7 1393 115
XM_011537502.2 XP_011535804.1 6 - 7 1393 115
XM_011537504.2 XP_011535806.1 6 - 7 1393 115
XM_011537507.2 XP_011535809.1 6 - 7 1393 115
XM_017020629.1 XP_016876118.1 6 - 7 1393 115
XM_017020631.1 XP_016876120.1 5 - 6 1201 115
XM_017020632.1 XP_016876121.1 5 - 6 1201 115
XM_017020636.1 XP_016876125.1 6 - 7 1393 115
GenBank mRNA AF209505.1 - 5 - 6 607 115
AK000630.1 - 6 - 7 1290 115
AK074415.1 - 3 - 4 419 115
AY358768.1 - 6 - 7 1171 115
BC012564.1 - 6 - 7 1355 115
BC068515.1 - 6 - 7 1391 115

View Full Product Details