The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031217.3 | NP_112494.3 | 13 - 14 | 2130 | 140 |
XM_017018379.1 | XP_016873868.1 | 13 - 14 | 2686 | 140 | |
XM_017018380.1 | XP_016873869.1 | 5 - 6 | 838 | 140 | |
GenBank mRNA | AK093850.1 | - | 9 - 10 | 1317 | 140 |
AK312576.1 | - | 13 - 14 | 2075 | 140 | |
AL136819.1 | - | 13 - 14 | 2077 | 140 | |
AY791349.1 | - | 13 - 14 | 2060 | 140 | |
BC048347.2 | - | 12 - 13 | 1995 | 140 |