The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002507.3 | NP_002498.1 | 1 - 2 | 193 | 88 |
GenBank mRNA | AK313654.1 | - | 1 - 2 | 193 | 88 |
BC050309.1 | - | 1 - 2 | 193 | 88 | |
DA115173.1 | - | 1 - 2 | 193 | 88 | |
M14764.1 | - | 1 - 2 | 181 | 88 |