The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015918.3 | NP_057002.2 | 1 - 2 | 49 | 64 |
NM_198202.1 | NP_937845.1 | 1 - 2 | 53 | 64 | |
XM_011538441.1 | XP_011536743.1 | 1 - 2 | 107 | 64 | |
GenBank mRNA | AF070660.1 | - | 1 - 2 | 36 | 64 |
AF117232.1 | - | 1 - 2 | 36 | 64 | |
AJ306296.1 | - | 1 - 2 | 36 | 64 | |
AK303144.1 | - | 1 - 2 | 34 | 64 | |
BC012505.1 | - | 1 - 2 | 36 | 64 | |
BI549018.1 | - | 1 - 2 | 58 | 64 | |
BI668578.1 | - | 1 - 2 | 57 | 64 | |
BU155588.1 | - | - | 42 | 64 |