Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCN1
Entrez Gene ID
10985
Gene Name
GCN1, eIF2 alpha kinase activator homolog
Gene Aliases
GCN1L, GCN1L1, PRIC295
Chromosome Location
Chr.12: 120127210 - 120194710 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006836.1 NP_006827.1 57 - 58 7905 62
GenBank mRNA BC021129.1 - 6 - 7 777 62
BC032335.1 - 6 - 7 732 62
BC040172.1 - 13 - 14 2066 62
BC046177.1 - 3 - 4 448 62
BC064346.1 - 5 - 6 621 62
BC113083.1 - 2 - 3 47 62
BC153881.1 - 57 - 58 7905 62
D86973.2 - 57 - 58 7905 62
U77700.1 - 35 - 36 5370 62
U88836.1 - 18 - 19 2863 62

View Full Product Details