Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCN1
Entrez Gene ID
10985
Gene Name
GCN1, eIF2 alpha kinase activator homolog
Gene Aliases
GCN1L, GCN1L1, PRIC295
Chromosome Location
Chr.12: 120127210 - 120194710 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006836.1 NP_006827.1 9 - 10 853 88
GenBank mRNA AK297271.1 - 9 - 10 843 88
U88837.1 - 8 - 9 832 88

View Full Product Details