Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LY9
- Entrez Gene ID
-
4063
- Gene Name
- lymphocyte antigen 9
- Gene Aliases
-
CD229, SLAMF3, hly9, mLY9
- Chromosome Location
-
Chr.1: 160796074 - 160828256 on Build GRCh38
View Full Product Details