Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSNAXIP1
Entrez Gene ID
55815
Gene Name
translin associated factor X interacting protein 1
Gene Aliases
TXI1
Chromosome Location
Chr.16: 67806573 - 67828068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288990.1 NP_001275919.1 7 - 8 1087 85
NM_001288991.1 NP_001275920.1 5 - 6 880 85
NM_001288992.1 NP_001275921.1 4 - 5 753 85
NM_001288993.1 NP_001275922.1 5 - 6 880 85
NM_001288994.1 NP_001275923.1 3 - 4 556 85
NM_018430.3 NP_060900.2 7 - 8 1276 85
XM_005256049.4 XP_005256106.1 7 - 8 1392 85
XM_011523230.2 XP_011521532.1 7 - 8 1392 85
XM_011523231.2 XP_011521533.1 7 - 8 1392 85
XM_011523232.2 XP_011521534.1 7 - 8 1392 85
XM_011523233.2 XP_011521535.1 7 - 8 1392 85
XM_011523234.2 XP_011521536.1 7 - 8 1392 85
XM_011523235.1 XP_011521537.1 7 - 8 1027 85
XM_011523236.2 XP_011521538.1 5 - 6 650 85
XM_011523237.2 XP_011521539.1 5 - 6 1152 85
XM_011523238.1 XP_011521540.1 4 - 5 565 85
XM_011523239.1 XP_011521541.1 5 - 6 566 85
XM_011523240.1 XP_011521542.1 6 - 7 752 85
XM_011523241.1 XP_011521543.1 5 - 6 840 85
XM_011523242.1 XP_011521544.1 4 - 5 693 85
XM_011523243.1 XP_011521545.1 4 - 5 647 85
XM_017023445.1 XP_016878934.1 5 - 6 650 85
XM_017023446.1 XP_016878935.1 4 - 5 565 85
XM_017023447.1 XP_016878936.1 5 - 6 650 85
XM_017023448.1 XP_016878937.1 4 - 5 565 85
XM_017023449.1 XP_016878938.1 5 - 6 650 85
XM_017023450.1 XP_016878939.1 6 - 7 752 85
XM_017023451.1 XP_016878940.1 6 - 7 938 85
XM_017023452.1 XP_016878941.1 4 - 5 693 85
XM_017023453.1 XP_016878942.1 5 - 6 840 85
XM_017023454.1 XP_016878943.1 4 - 5 693 85
XM_017023455.1 XP_016878944.1 4 - 5 693 85
GenBank mRNA AF132730.1 - 5 - 6 684 85
AK027245.1 - 7 - 8 994 85
AK301920.1 - 7 - 8 860 85
AK302300.1 - - 269 85
AK303841.1 - 5 - 6 670 85
BC111018.1 - 7 - 8 1030 85

View Full Product Details