The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005226.3 | NP_005217.2 | 2 - 2 | 2015 | 117 |
GenBank mRNA | AL832194.1 | - | 1 - 1 | 5369 | 117 |
BC060827.1 | - | 2 - 2 | 1977 | 117 | |
DR763542.1 | - | 1 - 1 | 191 | 117 |