Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP9B
Entrez Gene ID
374868
Gene Name
ATPase phospholipid transporting 9B (putative)
Gene Aliases
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location
Chr.18: 79069275 - 79378391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306085.1 NP_001293014.1 14 - 15 1660 105
NM_198531.4 NP_940933.3 14 - 15 1660 105
XM_011525963.2 XP_011524265.1 15 - 16 1826 105
XM_011525964.2 XP_011524266.1 14 - 15 1541 105
XM_011525965.2 XP_011524267.1 15 - 16 1822 105
XM_011525966.2 XP_011524268.1 14 - 15 1777 105
XM_011525971.2 XP_011524273.2 8 - 9 3884 105
XM_011525972.2 XP_011524274.1 9 - 10 901 105
XM_011525973.2 XP_011524275.1 7 - 8 675 105
XM_011525974.2 XP_011524276.1 7 - 8 697 105
XM_017025726.1 XP_016881215.1 15 - 16 1826 105
XM_017025727.1 XP_016881216.1 15 - 16 1826 105
XM_017025728.1 XP_016881217.1 15 - 16 1826 105
XM_017025729.1 XP_016881218.1 15 - 16 1826 105
XM_017025730.1 XP_016881219.1 15 - 16 1826 105
XM_017025731.1 XP_016881220.1 15 - 16 1826 105
XM_017025732.1 XP_016881221.1 15 - 16 1826 105
XM_017025733.1 XP_016881222.1 15 - 16 1826 105
XM_017025734.1 XP_016881223.1 15 - 16 1826 105
XM_017025735.1 XP_016881224.1 15 - 16 1826 105
XM_017025736.1 XP_016881225.1 15 - 16 1826 105
XM_017025737.1 XP_016881226.1 15 - 16 1826 105
XM_017025738.1 XP_016881227.1 11 - 12 1136 105
XM_017025739.1 XP_016881228.1 11 - 12 1154 105
XM_017025740.1 XP_016881229.1 10 - 11 1059 105
XM_017025741.1 XP_016881230.1 12 - 13 1321 105
XM_017025742.1 XP_016881231.1 15 - 16 1826 105
GenBank mRNA AK093704.1 - 11 - 12 1183 105
AK308291.1 - 14 - 15 1542 105
BC125219.1 - 14 - 15 1538 105
BC125220.1 - 14 - 15 1538 105
BX537860.1 - 12 - 13 1043 105
BX648948.1 - 2 - 3 235 105

View Full Product Details