Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP9B
Entrez Gene ID
374868
Gene Name
ATPase phospholipid transporting 9B (putative)
Gene Aliases
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location
Chr.18: 79069275 - 79378391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306085.1 NP_001293014.1 6 - 7 858 144
NM_198531.4 NP_940933.3 6 - 7 858 144
XM_011525963.2 XP_011524265.1 7 - 8 1024 144
XM_011525964.2 XP_011524266.1 6 - 7 739 144
XM_011525965.2 XP_011524267.1 7 - 8 1020 144
XM_011525966.2 XP_011524268.1 6 - 7 975 144
XM_017025726.1 XP_016881215.1 7 - 8 1024 144
XM_017025727.1 XP_016881216.1 7 - 8 1024 144
XM_017025728.1 XP_016881217.1 7 - 8 1024 144
XM_017025729.1 XP_016881218.1 7 - 8 1024 144
XM_017025730.1 XP_016881219.1 7 - 8 1024 144
XM_017025731.1 XP_016881220.1 7 - 8 1024 144
XM_017025732.1 XP_016881221.1 7 - 8 1024 144
XM_017025733.1 XP_016881222.1 7 - 8 1024 144
XM_017025734.1 XP_016881223.1 7 - 8 1024 144
XM_017025735.1 XP_016881224.1 7 - 8 1024 144
XM_017025736.1 XP_016881225.1 7 - 8 1024 144
XM_017025737.1 XP_016881226.1 7 - 8 1024 144
XM_017025738.1 XP_016881227.1 4 - 5 429 144
XM_017025739.1 XP_016881228.1 4 - 5 429 144
XM_017025740.1 XP_016881229.1 4 - 5 429 144
XM_017025741.1 XP_016881230.1 4 - 5 429 144
XM_017025742.1 XP_016881231.1 7 - 8 1024 144
GenBank mRNA AB373110.1 - 5 - 6 566 144
AK093704.1 - 5 - 6 553 144
AK097757.1 - 6 - 7 975 144
AK308291.1 - 6 - 7 740 144
BC125219.1 - 6 - 7 736 144
BC125220.1 - 6 - 7 736 144
BX537860.1 - 3 - 4 209 144

View Full Product Details