Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRB1
Entrez Gene ID
2560
Gene Name
gamma-aminobutyric acid type A receptor beta1 subunit
Gene Aliases
-
Chromosome Location
Chr.4: 47031278 - 47438409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000812.3 NP_000803.2 5 - 6 921 78
GenBank mRNA AK225167.1 - 5 - 6 598 78
AK293691.1 - 4 - 5 498 78
AK312720.1 - 5 - 6 567 78
BC022449.1 - 5 - 6 625 78
KJ534841.1 - 5 - 6 547 78
KJ535054.1 - 4 - 5 479 78
X14767.1 - 5 - 6 578 78

View Full Product Details